Learn More
Cayman Chemical ANTIBODY CDPPB 1mg
SDP
Supplier:  Cayman Chemical 341321 MG
Encompass_Preferred
A positive allosteric modulator of mGluR5; potentiates glutamate-induced increases in intracellular calcium in CHO cells expressing mGluR5 (EC50s = 10 and 20 nM for the human and rat receptor, respectively); inhibits amphetamine-induced PPI disruptions in rats at 30 mg/kg, s.c.
Catalog No. 50-294-6728