Learn More
Cayman Chemical ANTIBODY -Cedrol 100mg
SDP
Supplier:  Cayman Chemical 25769100 MG
Encompass_Preferred
A sesquiterpene alcohol; inhibits the growth of L. sulphureus, G. trabeum, L. betulina, and T. versicolor wood decay fungi at 100 μg/ml; inhibits CYP2B6 and CYP3A4 (Kis = 0.9 and 3.4 μM, respectively); prevents hair follicle dystrophy and reduces hair loss in a mouse model of cyclophosphamide-induced alopecia at 200 mg/kg; loaded nanostructured lipid particles reduce compound 48/80-induced peritoneal mast cell degranulation in mice
Catalog No. 50-293-5865