Learn More
Cayman Chemical ANTIBODY CemEC1 5mg
SDP
Supplier:  Cayman Chemical 199565 MG
Encompass_Preferred
A selective inhibitor of TAF1 bromodomain 2 (Kd = 1.8 µM; IC50 = 0.9 µM); synergizes with (+)-JQ1 to inhibit the proliferation of THP-1 and H23 lung adenocarcinoma cells
Catalog No. 50-298-1447