Learn More
Cayman Chemical ANTIBODY CGI1746 1mg
SDP
Supplier:  Cayman Chemical 184141 MG
Encompass_Preferred
A potent, selective inhibitor of BTK (IC50 = 1.9 nM); prevents BCR-mediated B lymphocyte proliferation; reduces cytokine levels within joints and ameliorates symptoms in a mouse model of autoantibody-induced arthritis
Catalog No. 50-296-5424