Learn More
Cayman Chemical ANTIBODY CHIR98014 10mg
SDP
Supplier:  Cayman Chemical 1557810 MG
Encompass_Preferred
A reversible, cell-permeable inhibitor of GSK3α and GSK3β (IC50 = 0.65 and 0.58 nM, respectively); stimulates glycogen synthase in cells (EC50 = 106 nM), potentiates insulin-dependent glucose transport in isolated muscle strips, and improves glucose disposal in diabetic animals
Catalog No. 50-297-1159