Learn More
Cayman Chemical ANTIBODY CHM-1 10mg
SDP
Supplier:  Cayman Chemical 2764710 MG
Encompass_Preferred
An inhibitor of tubulin polymerization (IC50 = 0.68 µM); inhibits chochicine tubulin binding at 5 µM; inhibits the growth of K562, NCI H226, HCT116, OVCAR-3, RXF 393L, SK-MEL-5, SF-268, and SF-295 cancer cells (mean GI50 = 130 nM); induces apoptosis in HA22T hepatocellular carcinoma cells; reduces tumor volume and increases survival in an HA22T mouse xenograft model at 10 mg/kg
Catalog No. 50-293-9877