Learn More
Cayman Chemical ANTIBODY CK-636 25mg
SDP
Supplier:  Cayman Chemical 1624325 MG
Encompass_Preferred
A cell-permeable inhibitor of Arp2/3 complex action that inhibits actin polymerization in vitro, using human, bovine and fission yeast proteins (IC50s = 4, 32, and 24 µM, respectively); prevents the formation of actin filament comet tails by Listeria in infected SKOV3 cells (IC50 = 22 µM)
Catalog No. 50-297-2989