Learn More
Cayman Chemical ANTIBODY CK-869 25mg
SDP
Supplier:  Cayman Chemical 1525125 MG
Encompass_Preferred
An inhibitor of actin polymerization via inhibition of the Arp2/3 complex; inhibits comet tail formation by L. monocytogenes (IC50 = 7 µM); impairs cell adhesion and spreading
Catalog No. 50-296-8829