Learn More
Cayman Chemical ANTIBODY CL 82 198 50mg
SDP
Supplier:  Cayman Chemical 1692850 MG
Encompass_Preferred
A selective inhibitor of human collagenase-3, also known as MMP-13, producing 89% inhibition at 10 UG/ml; used to evaluate the role of MMP-13 in diverse processes
Catalog No. 50-296-7119