Learn More
Cayman Chemical ANTIBODY ClopyralId 100mg
SDP
Supplier:  Cayman Chemical 24229100 MG
Encompass_Preferred
An herbicide; selectively inhibits the growth of the broadleaf weed L. palustris over a panel of 17 New Zealand native non-broadleaf plant species when applied at 1.5 mg/L; does not inhibit growth of human pluripotent stem cells (IC50 = >100 μM) or decrease nuclear translocation of the transcription factor SOX17
Catalog No. 50-293-3279