Learn More
Cayman Chemical ANTIBODY CMPF-d5 500UG
SDP
Supplier:  Cayman Chemical 9001871500 UG
Encompass_Preferred
An internal standard for the quantification of CMPF by GC- or LC-MS
Catalog No. 50-289-3354