Learn More
Cayman Chemical ANTIBODY Compound E 1mg
SDP
Supplier:  Cayman Chemical 155791 MG
Encompass_Preferred
A potent, cell-permeable, and selective inhibitor of γ-secretase, blocking the cleavage of both APP and Notch CTFs with IC50 values of ~0.3 nM; induces neuronal differentiation, impairs ovarian folliculogenesis, and suppresses thymocyte development by preventing Notch activation by γ-secretase
Catalog No. 50-296-9698