Learn More
Abcam Antibody Concentration and Clean Up Kit for Late x & Europium, 4UNIT
Supplier:  Abcam AB2699654UNIT
Encompass_Preferred
The Antibody Concentration and Clean Up Kit for Latex & Europium kit (ab269965) removes small molecule buffer components from the antibody or protein prior to use in the conjugation reaction.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-265-6248