Learn More
Cayman Chemical ANTIBODY CorIlagIn 10mg
SDP
Supplier:  Cayman Chemical 1183210 MG
Encompass_Preferred
A polyphenol and hydrolyzable tannin that inhibits squalene epoxidase (IC50 = 4.0 µM), a key enzyme in cholesterol synthesis; has various anti-inflammatory and anti-cancer effects
Catalog No. 50-295-2282