Learn More
Cayman Chemical ANTIBODY CP 346 086 1mg
SDP
Supplier:  Cayman Chemical 391561 MG
Encompass_Preferred
An MTTP inhibitor (IC50 = 2 nM); inhibits ApoB and triglyceride secretion in HepG2 cells (IC50 = 2.6 nM for both); lowers plasma triglyceride levels, as well as plasma total, VLDL, and LDL cholesterol levels in rats at 10 mg/kg
Catalog No. 50-291-6182