Learn More
Cayman Chemical ANTIBODY CP 640 186 5mg
SDP
Supplier:  Cayman Chemical 176915 MG
Encompass_Preferred
An ACC inhibitor (IC50s = 53 and 61 nM for rat liver ACC1 and rat skeletal muscle ACC2, respectively); lowers malonyl-CoA levels and inhibits hepatic fatty acid and triglyceride synthesis both in vitro and in vivo
Catalog No. 50-298-0585