Learn More
Cayman Chemical ANTIBODY CP 673 451 5mg
SDP
Supplier:  Cayman Chemical 191705 MG
Encompass_Preferred
A selective inhibitor of PDGFRβ and PDGFRα kinases (IC50s = 1 and 10 nM, respectively); antiangiogenic and antitumor activity in several in vivo tumor models
Catalog No. 50-298-4693