Learn More
Cayman Chemical ANTIBODY CP 724 714 10mg
SDP
Supplier:  Cayman Chemical 1917210 MG
Encompass_Preferred
A selective inhibitor of HER2/ErbB2 (IC50 = 10 nM); inhibits the proliferation of ErbB2-amplified cells, including BT474 and SK-BR-3 (IC50s = 0.25 and 0.95 μM, respectively) and demonstrates antitumor activity in various human tumor xenograft models
Catalog No. 50-298-4931