Learn More
Cayman Chemical ANTIBODY CPI-613 10mg
SDP
Supplier:  Cayman Chemical 1698110 MG
Encompass_Preferred
A lipoic acid analog that inhibits α-ketoglutarate dehydrogenase, particularly in tumor cells; induces a strong mitochondrial burst of reactive oxygen species at 60-240 µM, resulting in cell death; demonstrates both in vitro and in vivo anti-tumor activity
Catalog No. 50-296-7620