Learn More
Cayman Chemical ANTIBODY CPI-637 10mg
SDP
Supplier:  Cayman Chemical 2350010 MG
Encompass_Preferred
An inhibitor of CBP/EP300 bromodomains (IC50s = 30 and 51 nM in a TR-FRET assay); selective for CBP and EP300 over the BET family of bromodomains (IC50s = >10 μM); is active at bromodomain BRD9 (IC50 = 0.73 μM); inhibits CBP (IC50 = 0.3 μM in a BRET assay) and inhibits MYC expression (EC50 = 0.6 μM) in AMO-1 cells
Catalog No. 50-299-4631