Learn More
Cayman Chemical ANTIBODY Cu-ATSM 5mg
SDP
Supplier:  Cayman Chemical 171225 MG
Encompass_Preferred
A copper-containing compound with diverse biological activities; inhibits ferroptotic cell death induced by RSL3, erastin, or iron(II) (EC50s = 134, 169, and 171 nM, respectively), as well as RSL3-, erastin-, or iron-induced lipid peroxidation in N27 rat mesencephalic cells at 1 µM; increases SOD1G37R protein levels, metalation, and activity in the spinal cord in the SOD1G37R transgenic mouse model of ALS at 30 mg/kg per day; increases the number of α-motor neurons and reduces oxidatively modified proteins in the spinal cord, as well as increases the latency to fall in the rotarod test in the same ALS mouse model; versions containing positron-emitting copper isotopes have been used in PET imaging applications for selective labeling of hypoxic tissue
Catalog No. 50-297-9903