Learn More
Cayman Chemical ANTIBODY CU-CPT9b 1mg
SDP
Supplier:  Cayman Chemical 287231 MG
Encompass_Preferred
A TLR8 antagonist; inhibits R-848-induced activation of NF-ĸB in TLR8-overexpressing HEK-Blue cells (IC50 = 0.7 nM)
Catalog No. 50-293-7152