Learn More
Cayman Chemical ANTIBODY CV-6209-d5 500UG
SDP
Supplier:  Cayman Chemical 9002895500 UG
Encompass_Preferred
An internal standard for the quantification of CV-6209 by GC- or LC-MS
Catalog No. 50-289-5868