Learn More
Cayman Chemical ANTIBODY CVT-10216 1mg
SDP
Supplier:  Cayman Chemical 183181 MG
Encompass_Preferred
A selective, reversible inhibitor of ALDH2 (IC50 = 29 nM); reduces excessive alcohol drinking in alcohol-preferring rats, prevents self-administration of alcohol in Long-Evans rats, and produces anxiolytic effects
Catalog No. 50-296-7600