Learn More
Cayman Chemical ANTIBODY CX-516 25mg
SDP
Supplier:  Cayman Chemical 1493625 MG
Encompass_Preferred
An AMPA receptor modulator; increases the mean open time of AMPA receptors which increases the amplitude of trisynaptic responses to presynaptic stimulation in rat hippocampal slices; accelerates acquisition of a conditioned fear response in rats from 10-50 mg/kg; enhances performance in a spatial DNMS test in rats; reverses dominance between the initially dominant and submissive rat pairs in the rat reduction of submissive behavior model
Catalog No. 50-296-9085