Learn More
Cayman Chemical ANTIBODY Cy3-SE 1mg
SDP
Supplier:  Cayman Chemical 303931 MG
Encompass_Preferred
An amine-reactive fluorescent probe; ex/em = 550/570 nm, respectively; has commonly been used to label nucleic acids
Catalog No. 50-299-0802