Learn More
Cayman Chemical ANTIBODY CyazofamId 100mg
SDP
Supplier:  Cayman Chemical 24044100 MG
Encompass_Preferred
A fungicide; active against several isolates of P. infestans, as well as P. sojae, P. aphanidermatum, P. paddicum, P. sylvaticum, and A. cochlioides (EC50s = 0.008-0.03, 0.2, 0.02, 0.2, 0.2, and 0.2 UG/ml, respectively); selectively inhibits P. spinosum mitochondrial complex III activity over B. cinerea, S. cerevisiae, potato tuber, and rat liver mitochondrial complex III activities at 10 µM; protects against P. brassicae-induced root hair infections in cabbage when applied to dry soil at 10 mg/kg
Catalog No. 50-293-3247