Learn More
Cayman Chemical ANTIBODY CYC-116 5mg
SDP
Supplier:  Cayman Chemical 212055 MG
Encompass_Preferred
A potent Aurora kinase inhibitor (Kis = 8.0 and 9.2 nM for Aurora A and B kinases, respectively); selective over against a panel of CDKs and other kinases, with the exception of Flt-3 (Ki = 44 nM); exhibits broad-spectrum antiproliferative activity against a panel of human cancer cell lines (mean IC50 = 0.54 µM); orally bioavailable
Catalog No. 50-298-1243