Learn More
Cayman Chemical ANTIBODY CYM 50308 25mg
SDP
Supplier:  Cayman Chemical 1466725 MG
Encompass_Preferred
A novel agonist of S1P4 (EC50 = 56 nM) that displays 37-fold selectivity against S1P5 (EC50 = 2.1 μM) and no appreciable activity over the S1P1-3 subtypes at concentrations up to 25 μM
Catalog No. 50-296-8955