Learn More
Cayman Chemical ANTIBODY CYM 5442 1mg
SDP
Supplier:  Cayman Chemical 169251 MG
Encompass_Preferred
A selective SIP1 agonist (EC50 = 1.35 nM); activates S1P1-mediated p42/p44 MAPK phosphorylation in CHO-K1 cells transfected with S1P1 (EC50 = 46 nM); induces S1P1-dependent lymphopenia in mice, decreasing B and T cells by 65 and 85%, respectively, at 50 nM
Catalog No. 50-298-0504