Learn More
Cayman Chemical ANTIBODY CyromazIn 25mg
SDP
Supplier:  Cayman Chemical 2404125 MG
Encompass_Preferred
A triazine insecticide; completely inhibits larval development of the Australian sheep blow fly (0.5 ppm in rearing medium); reduces egg production by adult blow flies and subsequent larval development (1-20 μg/ml in drinking water); reduces proliferation of the house fly in chicken manure
Catalog No. 50-293-4153