Learn More
Cayman Chemical ANTIBODY CYT387 10mg
SDP
Supplier:  Cayman Chemical 1624610 MG
Encompass_Preferred
An ATP-competitive inhibitor of JAK1 and JAK2 (IC50s = 11 and 18 nM, respectively); causes growth suppression and apoptosis in JAK2-dependent hematopoietic cell lines; is efficacious in a mouse model of JAK2V617F-dependent myeloproliferative neoplasms
Catalog No. 50-297-0321