Learn More
Cayman Chemical ANTIBODY CYT997 5mg
SDP
Supplier:  Cayman Chemical 183955 MG
Encompass_Preferred
An inhibitor of microtubule polymerization (IC50 = ~3 µM); induces cell cycle arrest at the G2/M phase; cytotoxic against a panel of cancer cell lines (IC50s = 0.009-0.101 µM); increases HUVEC monolayer permeability (IC50 = ~0.08 µM); decreases liver metastasis in a murine DMH-induced colon cancer model at 5, 10, and 15 mg/kg
Catalog No. 50-298-0074