Learn More
Cayman Chemical ANTIBODY CZC-8004 5mg
SDP
Supplier:  Cayman Chemical 167185 MG
Encompass_Preferred
An aminopyrimidine that binds a range of tyrosine kinases, including ABL, BTK, FAK, FER, JAK1, SRC, SYK, TEC, TNK1, TYK2, and YES
Catalog No. 50-298-0271