Learn More
Cayman Chemical ANTIBODY D-Xylulose 1mg
SDP
Supplier:  Cayman Chemical 208301 MG
Encompass_Preferred
A ketopentose that is converted from xylitol in the glucoronate-xylulose pathway
Catalog No. 50-298-4908