Learn More
Cayman Chemical ANTIBODY D13 5mg
SDP
Supplier:  Cayman Chemical 298185 MG
Encompass_Preferred
An acylhydrazone antifungal; active against C. neoformans in vitro (MIC80 = 0.06 μg/ml); increases survival in mouse models of C. neoformans, C. albicans, or A. fumigatus infection at 20 mg/kg per day, p.o.
Catalog No. 50-298-8391