Learn More
Cayman Chemical ANTIBODY DabIgatrn 5mg
SDP
Supplier:  Cayman Chemical 171335 MG
Encompass_Preferred
A thrombin inhibitor (Ki = 0.0045 µM); an active metabolite dabigatran etexilate; an inhibitor of trypsin (Ki = 0.0503 µM); selective for thrombin and trypsin over plasmin, Factor Xa, activated protein C, and tissue plasminogen activator (tPA; Kis = 1.695, 3.76, 20.93, and 45.36 µM, respectively)
Catalog No. 50-297-9981