Learn More
Cayman Chemical ANTIBODY DAR-4M 1mg
SDP
Supplier:  Cayman Chemical 278971 MG
Encompass_Preferred
A fluorescent probe for the detection of NO; fluorescence intensity of DAR-4M increases with increasing NO concentration, however, the fluorescent yield can be increased further in the presence of both NO and other RNS, but not in the presence of non-NO RNS alone; upon oxidation, it is converted to DAR-4M T, which displays abs/em = 554/572 nm, respectively
Catalog No. 50-294-1606