Learn More
Cayman Chemical ANTIBODY DB07268 1mg
SDP
Supplier:  Cayman Chemical 222571 MG
Encompass_Preferred
A potent inhibitor of JNK1 (IC50 = 9 nM); binds to the ATP site of JNK1; selective for JNK1 over P38, ERK2, AKT1, CHK1, and PAK4 among others
Catalog No. 50-298-6179