Learn More
Cayman Chemical ANTIBODY DBZ 10mg
SDP
Supplier:  Cayman Chemical 1462710 MG
Encompass_Preferred
A γ-secretase inhibitor that blocks Notch processing (IC50 = 1.7 nM); reduces Aβ40 levels by 71% in an APP transgenic mouse model within 4 hours after a single 100 μM/kg dose
Catalog No. 50-297-2921