Learn More
Cayman Chemical ANTIBODY DeracoxIb 25mg
SDP
Supplier:  Cayman Chemical 2737125 MG
Encompass_Preferred
An NSAID and selective COX-2 inhibitor (IC50s = 0.63 and 23 μM for COX-2 and COX-1, respectively); increases weight bearing and decreases lameness and joint effusion in a canine model of urate crystal-induced intraarticular synovitis; induces less lesion formation in the gastric mucosa of healthy canines than aspirin at 1.5 mg/kg
Catalog No. 50-293-7281