Learn More
Cayman Chemical ANTIBODY Derquntel 25mg
SDP
Supplier:  Cayman Chemical 1944125 MG
Encompass_Preferred
An anthelmintic spiroindole that acts as a nicotinic acetylcholine receptor antagonist, causing flaccid paralysis and expulsion of nematodes
Catalog No. 50-298-2908