Learn More
Cayman Chemical ANTIBODY DIeldrIn 25mg
SDP
Supplier:  Cayman Chemical 2404325 MG
Encompass_Preferred
An organochlorine insecticide; increases cell proliferation and the estrogen receptor transactivation gene response in MCF-7 cells (5 µM); inhibits R1881-induced androgen activity in CHO cells (20 µM); LD50 = 46 mg/kg in rats
Catalog No. 50-293-4117