Learn More
Cayman Chemical ANTIBODY DIflunIsal 10g
SDP
Supplier:  Cayman Chemical 1918710 G
Encompass_Preferred
An NSAID that inhibits both COX-1 (IC50 = 113 µM) and COX-2 (IC50s = 8.2 and 134 µM for human whole blood assay and human-modified whole blood assays, respectively)
Catalog No. 50-298-1866