Learn More
Cayman Chemical ANTIBODY DIFMUP 1mg
SDP
Supplier:  Cayman Chemical 355001 MG
Encompass_Preferred
A fluorogenic substrate for phosphatases; upon enzymatic cleavage by phosphatases, 6,8-difluoro-7-hydroxy-4-methylcoumarin is released, which has an abs/em maxima of 358/455 nm, respectively, and its fluorescence can be used to quantify phosphatase activity; has been used to detect alkaline phosphatase in isolated rat tibia bone tissue
Catalog No. 50-291-0159