Learn More
Cayman Chemical ANTIBODY DIl 25mg
SDP
Supplier:  Cayman Chemical 3042325 MG
Encompass_Preferred
A fluorescent neuronal tracer that labels cell bodies, axons, dendrites, and dendritic spines; has been used as an anterograde tracer to study neuronal development in vivo and as a retrograde tracer to label hippocampal neuronal connections in fixed postmortem brain tissue; has been used as a long-term plasma membrane label in live cells; ex/em = 549/565 nm, respectively
Catalog No. 50-299-0830