Learn More
Novus Biologicals Antibody Diluent 2, 20 mL
SDP
Supplier:  Novus Biologicals 042203
Encompass
Antibody Diluent 2 (20 mL) for use on Jess, Abby, Wes, Peggy Sue and Sally Sue systems. We strongly recommends use of the Antibody Diluent for dilution of primary and secondary antibodies as it has been proven superior to any other buffer system tested.
Catalog No. NC0945078