Learn More
Cayman Chemical ANTIBODY DImInutol 1mg
SDP
Supplier:  Cayman Chemical 182371 MG
Encompass_Preferred
A cell-permeable purine derivative that inhibits the NADP-dependent oxidoreductase, NQO1 (Ki = 1.72 µM), to depolymerize microtubules and disrupt mitosis
Catalog No. 50-296-7262