Learn More
Cayman Chemical ANTIBODY DM1-SMe 1mg
SDP
Supplier:  Cayman Chemical 224771 MG
Encompass_Preferred
A mertansine analog; has in vitro activity against the PPTP panel of cancer cell lines (IC50s = 0.002 to >3 nM)
Catalog No. 50-296-4120