Learn More
Cayman Chemical ANTIBODY DMNQ 25mg
SDP
Supplier:  Cayman Chemical 1957125 MG
Encompass_Preferred
A 1,4-naphthoquinone that acts as a redox-cycling agent, typically increasing intracellular superoxide and hydrogen peroxide formation
Catalog No. 50-298-2426